Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H11orf1
Entrez Gene ID
101109995
Gene Name
chromosome 15 C11orf1 homolog
Gene Aliases
-
Chromosome Location
Chr.15: 24740809 - 24746295 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016017.4 XP_004016066.1 3 - 4 646 97

View Full Product Details