Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCL22
Entrez Gene ID
101123141
Gene Name
C-C motif chemokine ligand 22
Gene Aliases
-
Chromosome Location
Chr.14: 26958879 - 26966944 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015022.4 XP_004015071.1 1 - 2 91 64

View Full Product Details