Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM6
Entrez Gene ID
101103235
Gene Name
PR/SET domain 6
Gene Aliases
-
Chromosome Location
Chr.5: 29894006 - 30008506 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969991.1 XP_027825792.1 5 - 6 1366 78

View Full Product Details