Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIPPLY3
Entrez Gene ID
101109856
Gene Name
ripply transcriptional repressor 3
Gene Aliases
-
Chromosome Location
Chr.1: 292503944 - 292511454 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012108635.3 XP_011964025.2 3 - 4 237 96

View Full Product Details