Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1A
Entrez Gene ID
101103650
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases
-
Chromosome Location
Chr.7: 75328749 - 75374099 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971910.1 XP_027827711.1 4 - 5 1129 97

View Full Product Details