Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCJ
Entrez Gene ID
101118754
Gene Name
IQ motif containing J
Gene Aliases
-
Chromosome Location
Chr.1: 248901793 - 248963563 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963890.1 XP_027819691.1 2 - 3 133 112

View Full Product Details