Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LEP
Entrez Gene ID
443534
Gene Name
leptin
Gene Aliases
ob
Chromosome Location
Chr.4: 101297507 - 101302245 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968780.1 XP_027824581.1 1 - 2 166 72
GenBank mRNA FR688115.1 - 1 - 2 63 72
FR688116.1 - 1 - 2 63 72
FR688118.1 - 1 - 2 63 72
U62123.1 - 1 - 2 55 72
U84247.1 - 1 - 2 79 72

View Full Product Details