Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TARBP2
Entrez Gene ID
101102467
Gene Name
TARBP2 subunit of RISC loading complex
Gene Aliases
-
Chromosome Location
Chr.3: 142544694 - 142550271 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006291.4 XP_004006340.1 5 - 6 872 78
XM_012174155.3 XP_012029545.1 6 - 7 908 78
XM_012174157.2 XP_012029547.1 6 - 7 586 78
XM_012174158.3 XP_012029548.1 6 - 7 914 78

View Full Product Details