Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAIF1
Entrez Gene ID
101122260
Gene Name
nuclear apoptosis inducing factor 1
Gene Aliases
-
Chromosome Location
Chr.3: 8441942 - 8446971 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005612.4 XP_004005661.1 1 - 2 746 90

View Full Product Details