Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS14
Entrez Gene ID
101116494
Gene Name
mitochondrial ribosomal protein S14
Gene Aliases
-
Chromosome Location
Chr.12: 60282849 - 60359385 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013818.3 XP_004013867.1 2 - 3 262 85

View Full Product Details