Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF23
Entrez Gene ID
101118033
Gene Name
PHD finger protein 23
Gene Aliases
-
Chromosome Location
Chr.11: 36400986 - 36405418 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012640.3 XP_004012689.3 2 - 3 337 94
XM_027974997.1 XP_027830798.1 2 - 3 62 94

View Full Product Details