Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOK2
Entrez Gene ID
101119612
Gene Name
docking protein 2
Gene Aliases
-
Chromosome Location
Chr.2: 45914055 - 45918719 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004211.4 XP_004004260.3 1 - 2 184 101

View Full Product Details