Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTSZ
Entrez Gene ID
101101911
Gene Name
cathepsin Z
Gene Aliases
-
Chromosome Location
Chr.13: 59729445 - 59738941 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099797.2 XP_014955283.2 2 - 3 448 103

View Full Product Details