Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFA4L2
Entrez Gene ID
101123455
Gene Name
NDUFA4 mitochondrial complex associated like 2
Gene Aliases
-
Chromosome Location
Chr.3: 174015378 - 174017694 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006553.3 XP_004006602.1 2 - 3 205 76

View Full Product Details