Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NXN
Entrez Gene ID
101123570
Gene Name
nucleoredoxin
Gene Aliases
-
Chromosome Location
Chr.11: 41347672 - 41508258 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975166.1 XP_027830967.1 2 - 3 573 90
XM_027975167.1 XP_027830968.1 2 - 3 289 90

View Full Product Details