Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFHAS1
Entrez Gene ID
101102369
Gene Name
malignant fibrous histiocytoma amplified sequence 1
Gene Aliases
-
Chromosome Location
Chr.26: 27147032 - 27257899 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104587.2 XP_014960073.2 2 - 3 3188 71

View Full Product Details