Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLRN1
Entrez Gene ID
101118841
Gene Name
clarin 1
Gene Aliases
-
Chromosome Location
Chr.1: 258387580 - 258432666 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003254.4 XP_004003303.1 2 - 3 792 108

View Full Product Details