Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LSM11
Entrez Gene ID
101114109
Gene Name
LSM11, U7 small nuclear RNA associated
Gene Aliases
-
Chromosome Location
Chr.5: 72278563 - 72291162 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096165.2 XP_014951651.2 3 - 4 669 81

View Full Product Details