Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLDN
Entrez Gene ID
101105662
Gene Name
gliomedin
Gene Aliases
-
Chromosome Location
Chr.7: 60746254 - 60799570 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010614.4 XP_004010663.1 8 - 9 1030 62

View Full Product Details