Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY6G5C
Entrez Gene ID
101104100
Gene Name
lymphocyte antigen 6 family member G5C
Gene Aliases
-
Chromosome Location
Chr.20: 29459774 - 29462573 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958559.1 XP_027814360.1 2 - 3 183 67

View Full Product Details