Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG3
Entrez Gene ID
101105970
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 3
Gene Aliases
-
Chromosome Location
Chr.24: 22600925 - 22698532 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020842.4 XP_004020891.1 2 - 3 901 104

View Full Product Details