Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLCF1
Entrez Gene ID
101108788
Gene Name
cardiotrophin like cytokine factor 1
Gene Aliases
-
Chromosome Location
Chr.21: 47021386 - 47031076 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103211.2 XP_014958697.2 2 - 3 404 94

View Full Product Details