Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MNAT1
Entrez Gene ID
101104405
Gene Name
MNAT1 component of CDK activating kinase
Gene Aliases
-
Chromosome Location
Chr.7: 75752690 - 76037776 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010698.4 XP_004010747.1 5 - 6 691 90
XM_012181783.3 XP_012037173.1 5 - 6 691 90

View Full Product Details