Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPRED2
Entrez Gene ID
101117408
Gene Name
sprouty related EVH1 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 44850007 - 44970927 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966690.1 XP_027822491.1 2 - 3 1780 75
XM_027966691.1 XP_027822492.1 2 - 3 1789 75

View Full Product Details