Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A31
Entrez Gene ID
101109903
Gene Name
solute carrier family 22 member 31
Gene Aliases
-
Chromosome Location
Chr.14: 15057779 - 15062370 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977754.1 XP_027833555.1 4 - 5 732 97

View Full Product Details