Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIP7
Entrez Gene ID
101120941
Gene Name
nucleolar pre-rRNA processing protein NIP7
Gene Aliases
-
Chromosome Location
Chr.14: 39254856 - 39286617 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015098.4 XP_004015147.1 4 - 5 630 89
XM_012190037.3 XP_012045427.1 3 - 4 548 89

View Full Product Details