Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IST1
Entrez Gene ID
101102921
Gene Name
IST1 factor associated with ESCRT-III
Gene Aliases
-
Chromosome Location
Chr.14: 41989112 - 42024689 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015113.4 XP_004015162.1 3 - 4 393 78
XM_012190049.3 XP_012045439.1 3 - 4 393 78
XM_012190050.3 XP_012045440.1 3 - 4 397 78

View Full Product Details