Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZWINT
Entrez Gene ID
101115332
Gene Name
ZW10 interacting kinetochore protein
Gene Aliases
-
Chromosome Location
Chr.22: 3205414 - 3212611 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960303.1 XP_027816104.1 8 - 9 939 67

View Full Product Details