Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LSM1
Entrez Gene ID
101111937
Gene Name
LSM1 homolog, mRNA degradation associated
Gene Aliases
-
Chromosome Location
Chr.26: 36321667 - 36331431 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021778.3 XP_004021827.1 2 - 3 288 87

View Full Product Details