Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC8
Entrez Gene ID
101113724
Gene Name
thioredoxin domain containing 8
Gene Aliases
-
Chromosome Location
Chr.2: 13468670 - 13505330 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012113672.3 XP_011969062.1 2 - 3 126 74

View Full Product Details