Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VCPIP1
Entrez Gene ID
101122196
Gene Name
valosin containing protein interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.9: 47278197 - 47307188 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011714.4 XP_004011763.2 1 - 2 2708 94

View Full Product Details