Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101103379
Entrez Gene ID
101103379
Gene Name
protein NipSnap homolog 3A
Gene Aliases
NIPSNAP3A
Chromosome Location
Chr.2: 18931267 - 18942859 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964162.1 XP_027819963.1 5 - 6 794 77

View Full Product Details