Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL6
Entrez Gene ID
101102950
Gene Name
kelch like family member 6
Gene Aliases
-
Chromosome Location
Chr.1: 222074504 - 222131256 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003107.4 XP_004003156.1 2 - 3 550 58

View Full Product Details