Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP68
Entrez Gene ID
101117660
Gene Name
centrosomal protein 68
Gene Aliases
-
Chromosome Location
Chr.3: 45179197 - 45205995 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015094306.2 XP_014949792.2 5 - 6 2228 76
XM_015094307.2 XP_014949793.2 4 - 5 2143 76
XM_015094309.2 XP_014949795.2 5 - 6 2228 76
XM_015094311.2 XP_014949797.2 5 - 6 2228 76
XM_027966692.1 XP_027822493.1 5 - 6 2228 76

View Full Product Details