Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC7A6OS
Entrez Gene ID
101118809
Gene Name
solute carrier family 7 member 6 opposite strand
Gene Aliases
-
Chromosome Location
Chr.14: 38447341 - 38453709 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015090.4 XP_004015139.2 4 - 5 815 64

View Full Product Details