Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS24
Entrez Gene ID
101122388
Gene Name
mitochondrial ribosomal protein S24
Gene Aliases
-
Chromosome Location
Chr.19: 140940 - 151183 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018184.3 XP_004018233.1 2 - 3 246 77

View Full Product Details