Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFNB3
Entrez Gene ID
101103994
Gene Name
ephrin B3
Gene Aliases
-
Chromosome Location
Chr.11: 36016471 - 36022202 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012672.4 XP_004012721.1 1 - 2 195 67

View Full Product Details