Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TGFA
Entrez Gene ID
101106508
Gene Name
transforming growth factor alpha
Gene Aliases
TGF-a
Chromosome Location
Chr.3: 98811931 - 98889162 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966972.1 XP_027822773.1 2 - 3 269 95

View Full Product Details