Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD18
Entrez Gene ID
101122724
Gene Name
BTB domain containing 18
Gene Aliases
-
Chromosome Location
Chr.15: 87238963 - 87245074 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015101173.2 XP_014956659.1 1 - 2 124 70

View Full Product Details