Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ETHE1
Entrez Gene ID
101108154
Gene Name
ETHE1 persulfide dioxygenase
Gene Aliases
-
Chromosome Location
Chr.14: 55166141 - 55184435 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978406.1 XP_027834207.1 3 - 4 451 111

View Full Product Details