Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SAMD1
Entrez Gene ID
101118609
Gene Name
sterile alpha motif domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 9952410 - 9955383 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969506.1 XP_027825307.1 4 - 5 1506 70

View Full Product Details