Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTHRC1
Entrez Gene ID
101121943
Gene Name
collagen triple helix repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 81938244 - 81954252 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011798.4 XP_004011847.1 2 - 3 545 94

View Full Product Details