Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM20B
Entrez Gene ID
101120593
Gene Name
FAM20B glycosaminoglycan xylosylkinase
Gene Aliases
-
Chromosome Location
Chr.12: 64710186 - 64754738 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013832.4 XP_004013881.1 5 - 6 1351 98
XM_015099338.2 XP_014954824.1 5 - 6 945 98
XM_015099339.2 XP_014954825.1 5 - 6 2301 98
XM_027975910.1 XP_027831711.1 4 - 5 436 98

View Full Product Details