Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP5
Entrez Gene ID
101114099
Gene Name
THAP domain containing 5
Gene Aliases
-
Chromosome Location
Chr.4: 66218644 - 66224582 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007887.3 XP_004007936.2 1 - 2 217 111

View Full Product Details