Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISOC1
Entrez Gene ID
101102483
Gene Name
isochorismatase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 23651614 - 23672579 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012178174.3 XP_012033564.2 3 - 4 661 99
XM_027969967.1 XP_027825768.1 3 - 4 634 99

View Full Product Details