Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED6
Entrez Gene ID
101121367
Gene Name
transmembrane p24 trafficking protein 6
Gene Aliases
-
Chromosome Location
Chr.14: 39258211 - 39263434 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015100.4 XP_004015149.2 2 - 3 368 109

View Full Product Details