Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL27
Entrez Gene ID
101122495
Gene Name
interleukin 27
Gene Aliases
-
Chromosome Location
Chr.24: 26595860 - 26602230 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021214.4 XP_004021263.1 3 - 4 1319 66

View Full Product Details