Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBR1
Entrez Gene ID
101110826
Gene Name
T-box brain transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.2: 160237678 - 160247241 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964904.1 XP_027820705.1 4 - 5 1403 86

View Full Product Details