Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEBOX
Entrez Gene ID
101118969
Gene Name
SEBOX homeobox
Gene Aliases
-
Chromosome Location
Chr.11: 43520413 - 43523308 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015098533.2 XP_014954019.2 1 - 2 1595 58

View Full Product Details