Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS11F
Entrez Gene ID
101121519
Gene Name
transmembrane serine protease 11F
Gene Aliases
-
Chromosome Location
Chr.6: 92873039 - 92920020 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970749.1 XP_027826550.1 3 - 4 434 86

View Full Product Details